Assay Details
Target Gene Details
Entrez Gene ID: | 246788 |
Gene Name: | transient receptor potential cation channel, subfamily V, member 3 |
Gene Aliases: |
1110036I10Rik, AI644701, Nh, VRL3 |
Location: |
Chr.11:73267489-73300368 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trpv3 | NM_145099.2 | NP_659567.2 | ||
AB083654.1 | ||||
AF510316.1 | AAM33069.1 | |||
BC108984.1 | AAI08985.1 |
More Information
Set Membership: |
Intragenic Non-exonic |