Assay Details
Target Gene Details
Entrez Gene ID: | 19300 |
Gene Name: | ATP-binding cassette, sub-family D (ALD), member 4 |
Gene Aliases: |
P69r, P70R, Pxmp1l |
Location: |
Chr.12:84602531-84617479 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abcd4 | NM_008992.2 | NP_033018.2 | ||
AJ001166.1 | CAA04570.1 | |||
AK028420.1 | ||||
BB636710.1 | ||||
BC050102.1 | AAH50102.1 |
More Information
Set Membership: |
Intragenic Non-exonic |