Assay Details
Target Gene Details
Entrez Gene ID: | 215335 |
Gene Name: | solute carrier family 36 (proton/amino acid symporter), member 1 |
Gene Aliases: |
5830411H19Rik, AI839897, Pat1 |
Location: |
Chr.11:55204321-55236330 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc36a1 | NM_153139.4 | 10 | 1261 | NP_694779.3 |
AF453743.1 | 9 | 1247 | AAM80480.1 | |
AY211262.1 | 10 | 1231 | AAO37090.1 | |
BC138556.1 | 10 | 1225 | ||
BC138558.1 | 10 | 1225 |
More Information
Set Membership: |
Intragenic Exonic |