Assay Details
Target Gene Details
Entrez Gene ID: | 72469 |
Gene Name: | phospholipase C, delta 3 |
Gene Aliases: |
2610205J15Rik, mKIAA1964 |
Location: |
Chr.11:103069236-103101658 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plcd3 | NM_152813.3 | NP_690026.2 | ||
AK011892.1 | ||||
AK046669.1 | BAC32829.1 | |||
AK164809.1 | ||||
AK166977.1 | ||||
AK173311.1 | BAD32589.1 | |||
BC031392.1 | AAH31392.1 |
More Information
Set Membership: |
Intragenic Non-exonic |