Assay Details
Target Gene Details
Entrez Gene ID: | 192157 |
Gene Name: | suppressor of cytokine signaling 7 |
Gene Aliases: |
2310063P06Rik, C85125, Nap4 |
Location: |
Chr.11:97362296-97398542 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Socs7 | NM_138657.3 | 3 | 990 | NP_619598.1 |
AF424814.1 | 3 | 991 | AAL60516.1 | |
AK147580.1 | 3 | 1088 | BAE28006.1 | |
AK147650.1 | 3 | 1088 | BAE28049.1 |
More Information
Set Membership: |
Intragenic Exonic |