Assay Details
Target Gene Details
Entrez Gene ID: | 103655 |
Gene Name: | SEC14-like 4 (S. cerevisiae) |
Gene Aliases: |
AI256582, AW536553, SPF, TAP |
Location: |
Chr.11:4031378-4052571 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sec14l4 | NM_146013.1 | NP_666125.1 | ||
AK028954.1 | ||||
BC026948.1 | AAH26948.1 |