Assay Details
Target Gene Details
Entrez Gene ID: | 13650 |
Gene Name: | rhomboid family 1 (Drosophila) |
Gene Aliases: |
C16ORF8, Dist, Dist1, Egfr-rs, mKIAA4242 |
Location: |
Chr.11:32209584-32222331 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rhbdf1 | NM_001291818.1 | 4 | 503 | NP_001278747.1 |
NM_010117.2 | 4 | 503 | NP_034247.2 | |
AK144437.1 | 4 | 491 | ||
AK220566.1 | 3 | 1413 | BAD90543.1 | |
BC027346.1 | 4 | 448 | AAH27346.1 | |
BY733688.1 | 4 | 491 | ||
CN664154.1 | 1 | 84 |
More Information
Set Membership: |
Intragenic Exonic |