Assay Details
Target Gene Details
Entrez Gene ID: | 216578 |
Gene Name: | poly(A) polymerase gamma |
Gene Aliases: |
9630006B20Rik |
Location: |
Chr.11:23860145-23895296 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Papolg | NM_172555.2 | NP_766143.2 | ||
AK035810.1 | BAC29194.1 | |||
BC019635.1 | AAH19635.1 | |||
BC023030.1 | AAH23030.1 | |||
BC059271.1 | AAH59271.1 | |||
DQ103505.1 | AAZ80292.1 |
More Information
Set Membership: |
Intragenic Non-exonic |