Assay Details
Target Gene Details
Entrez Gene ID: | 327958 |
Gene Name: | PITPNM family member 3 |
Gene Aliases: |
A330068P14Rik, AI848332, Ackr6, Gm880 |
Location: |
Chr.11:72047528-72135889 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pitpnm3 | NM_001024927.2 | 12 | 1783 | NP_001020098.1 |
NM_001081641.1 | 11 | 1735 | NP_001075110.1 | |
AK147444.1 | 12 | 1740 | BAE27916.1 | |
AK147630.1 | 11 | 1568 | BAE28035.1 | |
BC112411.1 |
More Information
Set Membership: |
Intragenic Exonic |