Assay Details
Target Gene Details
Entrez Gene ID: | 18972 |
Gene Name: | polymerase (DNA directed), delta 2, regulatory subunit |
Gene Aliases: |
50kDa, p50, po1D2 |
Location: |
Chr.11:5872180-5878256 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pold2 | NM_008894.2 | 9 | 1205 | NP_032920.2 |
AK013901.1 | 9 | 1171 | ||
BC003342.1 | 9 | 1171 | AAH03342.1 | |
BM215242.2 | 3 | 397 | ||
CB205181.1 | 6 | 741 | ||
Z72486.1 | 9 | 1166 | CAA96567.1 |
More Information
Set Membership: |
Intragenic Exonic |