Assay Details
Target Gene Details
Entrez Gene ID: | 19175 |
Gene Name: | proteasome (prosome, macropain) subunit, beta type 6 |
Gene Aliases: |
Lmp19, Mpnd |
Location: |
Chr.11:70525357-70527858 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Psmb6 | NM_008946.4 | NP_032972.3 | ||
AK078437.1 | BAC37272.1 | |||
AK132042.1 | BAE20959.1 | |||
AK167123.1 | BAE39271.1 | |||
AK167227.1 | BAE39351.1 | |||
BC013897.1 | AAH13897.1 | |||
BY069663.1 | ||||
U13393.1 | AAA75375.1 |
More Information
Set Membership: |
Intragenic Non-exonic |