Assay Details
Target Gene Details
Entrez Gene ID: | 18972 |
Gene Name: | polymerase (DNA directed), delta 2, regulatory subunit |
Gene Aliases: |
50kDa, p50, po1D2 |
Location: |
Chr.11:5872180-5878256 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pold2 | NM_008894.2 | NP_032920.2 | ||
AK013901.1 | ||||
BC003342.1 | AAH03342.1 | |||
BG078511.2 | ||||
BY093766.1 | ||||
Z72486.1 | CAA96567.1 |
More Information
Set Membership: |
Intragenic Non-exonic |