Assay Details
Target Gene Details
Entrez Gene ID: | 72055 |
Gene Name: | solute carrier family 38, member 10 |
Gene Aliases: |
1810073N04Rik |
Location: |
Chr.11:120103951-120151351 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc38a10 | NM_001164798.1 | NP_001158270.1 | ||
NM_001164799.1 | NP_001158271.1 | |||
NM_001164800.1 | NP_001158272.1 | |||
NM_001164801.1 | NP_001158273.1 | |||
NM_001164802.1 | NP_001158274.1 | |||
NM_024249.5 | NP_077211.4 | |||
AK007977.1 | BAB25384.1 | |||
AK032245.1 | BAC27777.1 | |||
AK090050.1 | BAC41066.1 | |||
AK159948.1 | BAE35504.1 | |||
AK165672.1 | ||||
AK180057.1 | ||||
AK183661.1 | ||||
AK185748.1 | ||||
AK189102.1 | ||||
BC082300.1 | AAH82300.1 | |||
BC088811.1 | AAH88811.1 |
More Information
Set Membership: |
Intragenic Non-exonic |