Assay Details
Target Gene Details
Entrez Gene ID: | 18604 |
Gene Name: | pyruvate dehydrogenase kinase, isoenzyme 2 |
Gene Aliases: |
- |
Location: |
Chr.11:95026258-95041371 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pdk2 | NM_133667.2 | NP_598428.2 | ||
AF267660.1 | AAF72038.1 | |||
AK002647.1 | ||||
BC021764.1 | AAH21764.1 | |||
CK618575.1 |
More Information
Set Membership: |
Intragenic Non-exonic |