Assay Details
Target Gene Details
Entrez Gene ID: | 69900 |
Gene Name: | major facilitator superfamily domain containing 11 |
Gene Aliases: |
2600014M03Rik |
Location: |
Chr.11:116852283-116875660 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mfsd11 | NM_178620.3 | NP_848735.1 | ||
AF015191.1 | AAB92498.1 | |||
AK031384.1 | BAC27376.1 | |||
AK032043.1 | BAC27666.1 | |||
AK080006.1 | 3 | 1068 | ||
BC088988.1 | AAH88988.1 | |||
BC137970.1 | ||||
BC137971.1 |