Assay Details
Target Gene Details
Entrez Gene ID: | 16416 |
Gene Name: | integrin beta 3 |
Gene Aliases: |
CD61, GP3A, INGRB3 |
Location: |
Chr.11:104608000-104670490 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itgb3 | NM_016780.2 | NP_058060.2 | ||
AF026509.1 | AAB94086.1 | |||
AK156518.1 | ||||
AK157958.1 | BAE34283.1 | |||
BC125518.1 | ||||
BC125520.1 |
More Information
Set Membership: |
Intragenic Non-exonic |