Assay Details
Target Gene Details
Entrez Gene ID: | 67493 |
Gene Name: | methyltransferase like 16 |
Gene Aliases: |
2610100D03Rik, 2810013M15Rik, A830095F14Rik, AI846133, Mett10d |
Location: |
Chr.11:74770836-74818694 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mettl16 | NM_026197.3 | NP_080473.1 | ||
AK011780.1 | BAB27835.1 | |||
AK012739.1 | BAB28440.1 | |||
AK013306.1 | ||||
AK029045.1 | ||||
AK145022.1 | BAE26189.1 | |||
AK145662.1 | BAE26574.1 | |||
AK168798.1 | BAE40630.1 | |||
AK169540.1 | 2 | 194 | ||
BC049897.1 | AAH49897.1 | |||
BC054724.1 | AAH54724.1 |
More Information
Set Membership: |
Intragenic Non-exonic |