Assay Details
Target Gene Details
Entrez Gene ID: | 104601 |
Gene Name: | MYCBP associated protein |
Gene Aliases: |
4932408B01Rik, AMAP-1, AW125474 |
Location: |
Chr.11:94501347-94532388 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mycbpap | NM_170671.2 | 7 | 879 | NP_733771.2 |
AK029929.1 | 6 | 1055 | BAC26684.1 | |
BC027327.1 | 3 | 240 | AAH27327.1 |