Assay Details
Target Gene Details
Entrez Gene ID: | 23989 |
Gene Name: | mediator complex subunit 24 |
Gene Aliases: |
911GSE, AU040102, AW547152, D11Ertd307e, DRIP100, Gse2, Pparb2, Pparbp2, R75526, Thrap4, Trap100 |
Location: |
Chr.11:98704591-98729475 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Med24 | NM_011869.2 | NP_035999.2 | ||
AF126543.1 | AAD42776.1 | |||
AF483498.1 | AAL90772.1 | |||
AF483499.1 | AAL90773.1 | |||
AK020269.1 | BAB32051.1 | |||
AK154618.1 | BAE32717.1 | |||
AK158385.1 | ||||
AK219304.1 | ||||
BC005409.1 | AAH05409.1 | |||
BU612260.1 |
More Information
Set Membership: |
Intragenic Non-exonic |