Assay Details
Target Gene Details
Entrez Gene ID: | 94179 |
Gene Name: | keratin 23 |
Gene Aliases: |
CK23, Haik1, K23, Krt1-23 |
Location: |
Chr.11:99477973-99493110 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Krt23 | NM_033373.1 | NP_203537.1 | ||
AF102849.1 | AAK00051.1 | |||
AK036901.1 | BAC29630.1 | |||
AK036954.1 | BAC29649.1 | |||
AK145247.1 | BAE26325.1 | |||
BC019972.1 | AAH19972.1 |