Assay Details
Target Gene Details
Entrez Gene ID: | 327987 |
Gene Name: | mediator complex subunit 13 |
Gene Aliases: |
1110067M05Rik, D030023K18, Thrap1, Trap240 |
Location: |
Chr.11:86265715-86357525 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Med13 | NM_001080931.1 | 10 | 2033 | NP_001074400.1 |