Assay Details
Target Gene Details
Entrez Gene ID: | 171283 |
Gene Name: | hepatitis A virus cellular receptor 1 |
Gene Aliases: |
AI503787, KIM-1, TIM-1, Tim1, Timd1 |
Location: |
Chr.11:46739823-46779578 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Havcr1 | NM_001166631.1 | 4 | 600 | NP_001160103.1 |
NM_001166632.1 | 4 | 484 | NP_001160104.1 | |
NM_134248.2 | 3 | 439 | NP_599009.2 | |
AF399829.1 | 3 | 430 | AAL35774.1 | |
AF399830.1 | 3 | 430 | AAL35775.1 | |
AK133466.1 | 4 | 602 | BAE21672.1 | |
BC053400.1 | 4 | 492 | AAH53400.1 | |
CA551363.1 | 4 | 484 |