Assay Details
Target Gene Details
Entrez Gene ID: | 360013 |
Gene Name: | myosin XVIIIA |
Gene Aliases: |
MAJN, MyoPDZ, MysPDZ |
Location: |
Chr.11:77763251-77865988 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Myo18a | NM_001291212.1 | 7 | 1734 | NP_001278141.1 |
NM_001291213.1 | 7 | 1842 | NP_001278142.1 | |
NM_001291214.1 | 6 | 701 | NP_001278143.1 | |
NM_001291215.1 | 7 | 737 | NP_001278144.1 | |
NM_011586.3 | 6 | 1698 | NP_035716.1 | |
AB026497.1 | 6 | 1668 | BAA93660.1 | |
AK137574.1 | 6 | 689 | BAE23413.1 | |
AK147584.1 | 7 | 686 | BAE28009.1 | |
AK171342.1 | 6 | 698 | BAE42402.1 | |
BC138366.1 | 7 | 1676 |