Assay Details
Target Gene Details
Entrez Gene ID: | 16706 |
Gene Name: | kinase suppressor of ras 1 |
Gene Aliases: |
AW492498, B-KSR1, D11Bhm183e, D11Bhm184e, Ksr, RSU2, hb |
Location: |
Chr.11:79013440-79146554 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ksr1 | NM_013571.2 | NP_038599.1 | ||
AK036566.1 | ||||
AK164833.1 | ||||
U43585.1 | AAC52382.1 | |||
X81634.1 | CAA57288.1 |
More Information
Set Membership: |
Intragenic Non-exonic |