Assay Details
Target Gene Details
Entrez Gene ID: | 15254 |
Gene Name: | histidine triad nucleotide binding protein 1 |
Gene Aliases: |
AA673479, Hint, Ipk1, PKCI-1, PRKCNH1 |
Location: |
Chr.11:54866438-54870496 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hint1 | NM_008248.2 | NP_032274.1 | ||
AK002965.2 | BAB22484.1 | |||
AK012433.1 | BAB28235.1 | |||
BC070415.1 | AAH70415.1 | |||
BC080296.1 | AAH80296.1 | |||
U60001.1 | AAC71076.1 |
More Information
Set Membership: |
Intragenic Non-exonic |