Assay Details
Target Gene Details
Entrez Gene ID: | 70355 |
Gene Name: | G protein-coupled receptor, family C, group 5, member C |
Gene Aliases: |
1110028I06Rik, 3200002M13Rik, Raig3 |
Location: |
Chr.11:114851146-114872617 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gprc5c | NM_001110337.1 | NP_001103807.1 | ||
NM_001110338.1 | NP_001103808.1 | |||
NM_147217.3 | 1 | 79 | NP_671750.2 | |
AK128950.1 | ||||
AY079516.1 | AAL87525.1 | |||
BB871704.1 | 1 | 79 | ||
BC031439.1 | 1 | 56 | AAH31439.1 | |
BY003041.1 |
More Information
Set Membership: |
Intragenic Exonic |