Assay Details
Target Gene Details
Entrez Gene ID: | 16897 |
Gene Name: | lethal giant larvae homolog 1 (Drosophila) |
Gene Aliases: |
AI325176, Lgl1, Llglh, Mgl1 |
Location: |
Chr.11:60699670-60714188 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Llgl1 | NM_001159404.1 | 16 | 2306 | NP_001152876.1 |
NM_001159405.1 | 16 | 2306 | NP_001152877.1 | |
NM_008502.2 | 16 | 2300 | NP_032528.1 | |
AK084386.1 | 8 | 1176 | BAC39171.1 | |
AK159412.1 | 16 | 2172 | BAE35062.1 | |
BC043458.1 | 2 | 211 | AAH43458.1 | |
BC050913.1 | 16 | 2151 | AAH50913.1 | |
BC055399.1 | 16 | 2273 | AAH55399.1 | |
D16141.1 | 16 | 2150 | BAA03712.1 |
More Information
Set Membership: |
Intragenic Exonic |