Assay Details
Target Gene Details
Entrez Gene ID: | 217127 |
Gene Name: | K(lysine) acetyltransferase 7 |
Gene Aliases: |
Hbo1, Hboa, Myst2 |
Location: |
Chr.11:95271853-95310246 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kat7 | NM_001195003.1 | 8 | 1105 | NP_001181932.1 |
NM_001195004.1 | 7 | 1015 | NP_001181933.1 | |
NM_177619.3 | 6 | 838 | NP_808287.1 | |
AK147968.1 | 7 | 832 | BAE28257.1 | |
AK169458.1 | 6 | 936 | ||
AK170389.1 | 7 | 962 | BAE41763.1 | |
AK186978.1 | 3 | 190 | ||
BC048904.1 | 6 | 793 | AAH48904.1 | |
BC057102.1 | 7 | 943 | AAH57102.1 | |
DQ076247.1 | 8 | 1033 |
More Information
Set Membership: |
Intragenic Exonic |