Assay Details
Target Gene Details
Entrez Gene ID: | 360013 |
Gene Name: | myosin XVIIIA |
Gene Aliases: |
MAJN, MyoPDZ, MysPDZ |
Location: |
Chr.11:77763251-77865988 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Myo18a | NM_001291212.1 | NP_001278141.1 | ||
NM_001291213.1 | NP_001278142.1 | |||
NM_001291214.1 | NP_001278143.1 | |||
NM_001291215.1 | NP_001278144.1 | |||
NM_011586.3 | NP_035716.1 | |||
AB026497.1 | BAA93660.1 | |||
AK137574.1 | BAE23413.1 | |||
AK147584.1 | BAE28009.1 | |||
AK171342.1 | BAE42402.1 | |||
BC138366.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |