Assay Details
Target Gene Details
Entrez Gene ID: | 16416 |
Gene Name: | integrin beta 3 |
Gene Aliases: |
CD61, GP3A, INGRB3 |
Location: |
Chr.11:104608000-104670490 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itgb3 | NM_016780.2 | 9 | 1277 | NP_058060.2 |
AF026509.1 | 9 | 1221 | AAB94086.1 | |
AK156518.1 | 5 | 534 | ||
AK157958.1 | 9 | 1223 | BAE34283.1 | |
BC125518.1 | 9 | 1277 | ||
BC125520.1 | 9 | 1277 |
More Information
Set Membership: |
Intragenic Exonic |