Assay Details
Target Gene Details
Entrez Gene ID: | 214384 |
Gene Name: | myocardin |
Gene Aliases: |
BSAC2A, Srfcp |
Location: |
Chr.11:65173608-65269989 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Myocd | NM_145136.4 | NP_660118.3 | ||
NM_146386.3 | NP_666498.2 | |||
AF384055.2 | AAK71683.2 | |||
AF437877.1 | AAL30892.1 | |||
AK142216.1 | BAE24980.1 | |||
AY303755.1 | AAQ63841.1 | |||
BC141281.1 |
More Information
Set Membership: |
Intragenic Non-exonic |