Assay Details
Target Gene Details
Entrez Gene ID: | 19157 |
Gene Name: | cytohesin 1 |
Gene Aliases: |
CLM1, CTH-1, CYTIP, Pscd1, mKIAA4240 |
Location: |
Chr.11:118164166-118248592 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyth1 | NM_001112699.1 | 8 | 716 | NP_001106169.1 |
NM_001112700.1 | 8 | 656 | NP_001106170.1 | |
NM_011180.3 | 8 | 716 | NP_035310.2 | |
AB013464.1 | 8 | 637 | BAA33428.1 | |
AB035538.1 | 8 | 685 | BAB13509.1 | |
AF051337.1 | 7 | 584 | AAC71694.1 | |
AK158216.1 | 8 | 656 | BAE34408.1 | |
AK172296.1 | 8 | 661 | BAE42931.1 | |
AK220302.1 | 8 | 638 | BAD90227.1 | |
AY126516.1 | 8 | 685 | AAM95454.1 | |
BC057974.1 | 8 | 600 | AAH57974.1 |
More Information
Set Membership: |
Intragenic Exonic |