Assay Details
Target Gene Details
Entrez Gene ID: | 13649 |
Gene Name: | epidermal growth factor receptor |
Gene Aliases: |
9030024J15Rik, AI552599, Erbb, Errb1, Errp, Wa5, wa-2, wa2 |
Location: |
Chr.11:16752203-16913907 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 22 - Intron 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Egfr | NM_207655.2 | NP_997538.1 | ||
AF275367.1 | AAG24386.1 | |||
AK087861.1 | ||||
BQ553815.1 | ||||
U03425.1 | AAA17899.1 | |||
X78987.1 | CAA55587.1 |
More Information
Set Membership: |
Intragenic Non-exonic |