Assay Details
Target Gene Details
Entrez Gene ID: | 216871 |
Gene Name: | glycolipid transfer protein domain containing 2 |
Gene Aliases: |
C730027E14Rik |
Location: |
Chr.11:70519209-70520736 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gltpd2 | NM_146020.1 | 2 | 389 | NP_666132.1 |
AK050107.1 | 2 | 403 | BAC34069.1 | |
AK050212.1 | 2 | 403 | BAC34125.1 | |
BC025515.1 | 2 | 388 | AAH25515.1 | |
BC030735.1 | 2 | 389 | AAH30735.1 |
More Information
Set Membership: |
Intragenic Exonic |