Assay Details
Target Gene Details
Entrez Gene ID: | 14172 |
Gene Name: | fibroblast growth factor 18 |
Gene Aliases: |
D130055P09Rik, FGF-18 |
Location: |
Chr.11:33116970-33147400 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fgf18 | NM_008005.2 | NP_032031.1 | ||
NR_102395.1 | ||||
AB004639.1 | BAA31980.1 | |||
AF075291.1 | AAC62239.1 | |||
AF211187.1 | AAF22976.1 | |||
AK078245.1 |
More Information
Set Membership: |
Intragenic Non-exonic |