Assay Details
Target Gene Details
Entrez Gene ID: | 238011 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 7 |
Gene Aliases: |
Alk-SMase, Gm254 |
Location: |
Chr.11:118988108-118993270 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Enpp7 | NM_001030291.1 | NP_001025462.1 | ||
AK144474.1 | 5 | 1502 | BAE25908.1 | |
AK167678.1 | 5 | 1502 | BAE39727.1 | |
BC141245.1 | 5 | 1584 | ||
BC171967.1 | 5 | 1584 |