Assay Details
Target Gene Details
Entrez Gene ID: | 246746 |
Gene Name: | CD300 antigen like family member F |
Gene Aliases: |
CLIM1, CLM-1, CLM1, Digr2, F730004D16Rik, IREM1, IgSF13, LMIR3, Pigr3 |
Location: |
Chr.11:115116214-115134150 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cd300lf | NM_001169153.1 | NP_001162624.1 | ||
NM_145634.3 | NP_663609.2 | |||
AB292061.1 | ||||
AF251703.1 | AAM19096.1 | |||
AK089313.1 | BAC40838.1 | |||
AK154273.1 | BAE32481.1 | |||
AK155367.1 | BAE33222.1 | |||
AY214460.1 | AAO59487.1 | |||
AY457047.1 | AAR27938.1 | |||
BC057864.1 | AAH57864.1 | |||
JX073136.1 |
Target Gene Details
Entrez Gene ID: | 58222 |
Gene Name: | RAB37, member RAS oncogene family |
Gene Aliases: |
B230331O03Rik, B230354I04Rik |
Location: |
Chr.11:115091203-115162773 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rab37 | NM_001163753.1 | NP_001157225.1 | ||
AK045995.1 | ||||
AK046260.1 | BAC32661.1 | |||
AK134576.1 | ||||
AK138293.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |