Assay Details
Target Gene Details
Entrez Gene ID: | 13195 |
Gene Name: | dopa decarboxylase |
Gene Aliases: |
Aadc |
Location: |
Chr.11:11814101-11898144 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddc | NM_001190448.1 | 6 | 783 | NP_001177377.1 |
NM_016672.4 | 6 | 721 | NP_057881.1 | |
AF071068.1 | 5 | 654 | AAC25566.1 | |
AK011834.1 | 6 | 715 | ||
AK149141.1 | 6 | 715 | BAE28750.1 |
More Information
Set Membership: |
Intragenic Exonic |