Assay Details
Target Gene Details
Entrez Gene ID: | 217303 |
Gene Name: | CD300A antigen |
Gene Aliases: |
B230315M08Rik, Clm8, LMIR1, MAIR-I, MAIR-Ia, MMAC8, Pigr4, mcpir1 |
Location: |
Chr.11:114889149-114904651 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cd300a | NM_170758.3 | NP_739564.1 | ||
AB065156.1 | BAC22595.1 | |||
AB091765.1 | BAC77074.1 | |||
AB091766.1 | BAC77075.1 | |||
AB095675.1 | BAC80268.1 | |||
AK045869.1 | BAC32515.1 | |||
AK053160.1 | BAC35290.1 | |||
AY457054.1 | AAR27945.1 | |||
BC139296.1 | ||||
BC139297.1 |
More Information
Set Membership: |
Intragenic Non-exonic |