Assay Details
Target Gene Details
Entrez Gene ID: | 171212 |
Gene Name: | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 10 |
Gene Aliases: |
AU018154, C330012K04Rik, GalNAc-T10, GalNAc-T9, Galnt9, ppGaNTase |
Location: |
Chr.11:57622407-57787501 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Galnt10 | NM_134189.2 | 11 | 1694 | NP_598950.2 |
AK033515.1 | 10 | 1352 | BAC28334.1 | |
AK131155.1 | 11 | 1672 | BAD21405.1 | |
AK143714.1 | 10 | 1525 | ||
AK159088.1 | 11 | 1693 | BAE34806.1 | |
AK159362.1 | 11 | 1692 | BAE35020.1 | |
AK162424.1 | 11 | 1695 | ||
BC016585.1 | 6 | 806 | AAH16585.1 | |
BC060617.1 | 11 | 1677 | AAH60617.1 |
More Information
Set Membership: |
Intragenic Exonic |