Assay Details
Target Gene Details
Entrez Gene ID: | 68626 |
Gene Name: | elaC homolog 2 (E. coli) |
Gene Aliases: |
1110017O07Rik, D11Wsu80e, Hpc2 |
Location: |
Chr.11:64979023-65002076 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Elac2 | NM_023479.2 | 7 | 674 | NP_075968.2 |
AF308696.2 | 7 | 612 | AAG24918.2 | |
AK156730.1 | 7 | 675 | BAE33824.1 | |
AK172604.1 | 6 | 1043 | ||
BC048235.1 | 7 | 608 | AAH48235.1 |
More Information
Set Membership: |
Intragenic Exonic |