Assay Details
Target Gene Details
Entrez Gene ID: | 276770 |
Gene Name: | eukaryotic translation initiation factor 5A |
Gene Aliases: |
AA410058, D19Wsu54e, Eif4d, Eif5a1, eIF-4D, eIF-5A, eIF-5A-1, eIF-5A1 |
Location: |
Chr.11:69916712-69921958 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif5a | NM_001166589.1 | 1 | 56 | NP_001160061.1 |
NM_001166591.1 | 1 | 56 | NP_001160063.1 | |
NM_001166593.1 | NP_001160065.1 | |||
NM_181582.4 | 1 | 56 | NP_853613.1 | |
AK011306.1 | BAB27532.1 | |||
AY129323.1 | AAN17519.1 | |||
AY129325.1 | 1 | 56 | AAN17528.1 | |
AY129326.1 | 1 | 56 | AAN17529.1 | |
AY129327.1 | 1 | 56 | AAN17533.1 |
More Information
Set Membership: |
Intragenic Exonic |