Assay Details
Target Gene Details
Entrez Gene ID: | 20306 |
Gene Name: | chemokine (C-C motif) ligand 7 |
Gene Aliases: |
MCP-3, Scya7, fic, marc, mcp3 |
Location: |
Chr.11:82045712-82047525 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccl7 | NM_013654.3 | NP_038682.1 | ||
AK078827.1 | BAC37413.1 | |||
BC061126.1 | AAH61126.1 | |||
BY163435.1 | ||||
S71251.1 | AAB30997.1 |
More Information
Set Membership: |
Intragenic Non-exonic |