Assay Details
Target Gene Details
Entrez Gene ID: | 74203 |
Gene Name: | eukaryotic translation initiation factor 4E nuclear import factor 1 |
Gene Aliases: |
2610509L04Rik, A930019J01Rik, AA410001, AU021239, Clast4, D11Ertd166e |
Location: |
Chr.11:3201598-3244588 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif4enif1 | NM_001166547.1 | 6 | 957 | NP_001160019.1 |
NM_001166548.1 | 6 | 961 | NP_001160020.1 | |
NM_001166549.1 | 6 | 1065 | NP_001160021.1 | |
NM_023743.2 | 6 | 972 | NP_076232.2 | |
AB031388.1 | 6 | 793 | BAB11963.1 | |
AK041598.1 | 6 | 923 | BAC30999.1 | |
AK088425.1 | 1 | 59 | BAC40347.1 | |
AK090363.1 | 5 | 1425 | ||
AK128990.1 | 6 | 1063 | ||
AK165678.1 | 6 | 960 | ||
AK205705.1 | 1 | 138 | ||
BC033410.1 | 6 | 807 | AAH33410.1 |
More Information
Set Membership: |
Intragenic Exonic |