Assay Details
Target Gene Details
Entrez Gene ID: | 14710 |
Gene Name: | guanine nucleotide binding protein (G protein), gamma transducing activity polypeptide 2 |
Gene Aliases: |
AV096488 |
Location: |
Chr.11:95842219-95845734 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gngt2 | NM_001038664.2 | 4 | 481 | NP_001033753.2 |
NM_001284393.1 | 4 | 485 | NP_001271322.1 | |
NM_001284397.1 | 4 | 482 | NP_001271326.1 | |
NM_023121.2 | 4 | 466 | NP_075610.1 | |
AA028742.1 | 4 | 436 | ||
AA423571.1 | 4 | 355 | ||
AK010554.1 | 3 | 1495 | BAB27023.1 | |
BC061005.1 | 4 | 431 | AAH61005.1 | |
BE847751.1 | ||||
BE994683.1 | 1 | 106 | ||
BF470980.1 | 3 | 295 |
More Information
Set Membership: |
Intragenic Exonic |