Assay Details
Target Gene Details
Entrez Gene ID: | 14710 |
Gene Name: | guanine nucleotide binding protein (G protein), gamma transducing activity polypeptide 2 |
Gene Aliases: |
AV096488 |
Location: |
Chr.11:95842219-95845734 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gngt2 | NM_001038664.2 | NP_001033753.2 | ||
NM_001284393.1 | NP_001271322.1 | |||
NM_001284397.1 | NP_001271326.1 | |||
NM_023121.2 | NP_075610.1 | |||
AA028742.1 | ||||
AA423571.1 | ||||
AI154009.1 | ||||
AI159118.1 | ||||
AK010554.1 | 1 | 1132 | BAB27023.1 | |
BC061005.1 | AAH61005.1 | |||
BE847751.1 | ||||
BY117112.1 | ||||
W35863.1 |
More Information
Set Membership: |
Intragenic Non-exonic |