Assay Details
Target Gene Details
Entrez Gene ID: | 14257 |
Gene Name: | FMS-like tyrosine kinase 4 |
Gene Aliases: |
AI323512, Chy, Flt-4, VEGFR-3, VEGFR3 |
Location: |
Chr.11:49609679-49652739 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 22 - Intron 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Flt4 | NM_008029.3 | NP_032055.1 | ||
BC138346.1 | ||||
BC138348.1 | ||||
L07296.1 | AAA40077.1 |
More Information
Set Membership: |
Intragenic Non-exonic |