Assay Details
Target Gene Details
Entrez Gene ID: | 268390 |
Gene Name: | AHA1, activator of heat shock protein ATPase 2 |
Gene Aliases: |
- |
Location: |
Chr.11:23487882-23498039 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ahsa2 | NM_001290654.1 | NP_001277583.1 | ||
NM_001290655.1 | NP_001277584.1 | |||
NM_172391.4 | NP_765979.3 | |||
AK033696.1 | ||||
AK035941.1 | BAC29251.1 | |||
AK093945.1 | BAC04256.1 | |||
AK137116.1 | BAE23240.1 | |||
AK154196.1 | BAE32431.1 | |||
AK155265.1 | BAE33153.1 | |||
BC038397.1 | ||||
BC052829.1 | AAH52829.1 | |||
BC064012.1 | AAH64012.1 |
More Information
Set Membership: |
Intragenic Non-exonic |