Assay Details
Target Gene Details
Entrez Gene ID: | 72947 |
Gene Name: | 5-phosphohydroxy-L-lysine phospholyase |
Gene Aliases: |
2900006B13Rik, Agxt2l2 |
Location: |
Chr.11:51583983-51603269 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Phykpl | NM_028398.2 | 11 | 1299 | NP_082674.1 |
AK013489.1 | 11 | 1253 | ||
AK156866.1 | 11 | 1644 | ||
AK160557.1 | 11 | 1268 | BAE35872.1 | |
BC024461.1 | 11 | 1253 | AAH24461.1 |