Assay Details
Target Gene Details
Entrez Gene ID: | 11764 |
Gene Name: | adaptor protein complex AP-1, beta 1 subunit |
Gene Aliases: |
Adtb1, b2b1660Clo |
Location: |
Chr.11:4947521-5042794 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ap1b1 | NM_001243043.1 | 7 | 1227 | NP_001229972.1 |
NM_001243044.1 | 7 | 1030 | NP_001229973.1 | |
NM_007454.3 | 7 | 1030 | NP_031480.2 | |
AK034140.1 | 7 | 1230 | BAC28603.1 | |
AK155892.1 | 7 | 950 | BAE33487.1 | |
AK159273.1 | 7 | 921 | BAE34952.1 | |
AK160042.1 | 7 | 921 | BAE35584.1 | |
AK165747.1 | 7 | 947 | ||
AK170376.1 | 7 | 941 | ||
AK204765.1 | ||||
BC008513.1 | 7 | 843 | AAH08513.1 | |
CA315828.1 | 2 | 250 | ||
Y07919.1 | 7 | 885 | CAA69224.1 |
More Information
Set Membership: |
Intragenic Exonic |